Learn More
Gibco™ Human IL-6R (soluble) Recombinant Protein, PeproTech®
Description
PeproTech's CHO cell-derived Recombinant Human sIL-6 Receptor alpha is a 37.9 kDa glycoprotein corresponding to 339 amino acid residues of the extracellular domain of IL-6Ralpha. As a result of glycosylation, Recombinant Human sIL-6 Receptor alpha migrates with an apparent molecular mass of approximately 57-70 kDa by SDS-PAGE gel, under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P08887 |
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 3570 |
| Molecular Weight (g/mol) | 37.9 kDa |
| Name | Human IL-6 R (soluble) |
| Quantity | 100 μg |
| Source | CHO cells |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.