missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-6 Quantikine ELISA Kit
Human IL-6 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Specifications
| Assay Range | 3.12 to 300 pg/mL |
|---|---|
| Assay Sensitivity | 0.7 pg/mL |
| Sample Type | Cell Culture Supernatant, Serum, Plasma (EDTA, Citrate, Heparin), Plasma (EDTA, Citrate, Heparin), Serum |
Description
The Human IL-6 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-6 in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul).Specifications
| 3.12 to 300 pg/mL | |
| 0.7 pg/mL | |
| Cell Culture Supernatant, Serum, Plasma (EDTA, Citrate, Heparin), Plasma (EDTA, Citrate, Heparin), Serum |
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.