missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems Human IL-6 Quantikine ELISA Kit
Click to view available options
Quantity:
1 Kit
1 Pack (6 Plates)
Unit Size:
1 set
Beskrivning
The Quantikine Human IL-6 Immunoassay is a 4.5 hour solid phase immunoassay designed to measure human IL-6 in cell culture supernates, serum, plasma, and urine. It contains recombinant human IL-6 and antibodies raised against the recombinant protein. Natural human IL-6 showed dose-response curves that were parallel to the standard curves obtained using the Quantikine kit standards, indicating that this kit can be used to determine relative levels of natural human IL-6.
Specifikationer
Specifikationer
| Assay Range | 3.13 - 300 pg/mL |
| Assay Sensitivity | 0.626 pg/mL |
| Sample Type | Cell Culture Supernatant, Serum, Plasma (EDTA), Plasma (Heparin), Urine |
| Specificity | Natural and recombinant human IL-6. |
| Gene ID (Entrez) | 3569 |
| Quantity | 1 Pack (6 Plates) |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Synonym | B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A |
| Target | Human |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering