missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-6 Magnetic Luminex™ Performance Assay
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
- Magnetic format allows for easier wash steps
- Undergoes similar validation testing as Quantikine™ ELISA Kits
- Precise and Specific
- Requires a small sample volume (<50 μL)
- Run your assay in just 3-5 hours
- Mass-calibrated standards for consistent results with every new lot of material
- Enough reagents for one 96-well plate
Specifications
Specifications
| Product Type | Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform |
| Quantity | 1 Kit |
| Species | Human |
| Binding Target | IL-6 |
| Bead Type | Magnetic |
| Assay Sensitivity | 1.11 pg/mL |
| Assay Range | 7.00 to 4950 pg/mL |
| Gene ID (Entrez) | 3569 |
| Gene Symbol | IL6 |
| Sample Type | Cell Culture Supernatants, Serum, Plasma (EDTA, Heparin) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction