missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-6 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 29 publications
Brand: R&D Systems AF-206-NA
432.65 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IL-6 Polyclonal specifically detects IL-6 in Human samples. It is validated for Western Blot, Simple Western, Neutralization, Immunocytochemistry.
Specifications
| IL-6 | |
| Polyclonal | |
| Unconjugated | |
| P05231 | |
| IL6 | |
| E. coli-derived recombinant human IL-6 Pro29-Met212 Accession # P05231 | |
| 100 μg | |
| Primary | |
| Detects human IL-6 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse IL-6 is observed. | |
| Human | |
| IgG |
| Neutralization, Immunocytochemistry, Immunocytochemistry | |
| LYOPH | |
| Neutralization 15-45 ng/mL, Immunocytochemistry 5-15 ug/mL | |
| B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3569 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction