missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-6 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC2062G-025
This item is not returnable.
View return policy
Description
IL-6 Monoclonal specifically detects IL-6 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-6 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3569 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 903129 | |
| Intracellular Staining by Flow Cytometry 5 uL/10^6 cells | |
| Q75MH2 | |
| IL6 | |
| E. coli-derived recombinant human IL-6 Pro29-Met212 Accession # Q75MH2 | |
| 25 Tests | |
| Primary | |
| Detects human IL-6 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction