missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-5 High Sensitivity Magnetic Luminex™ Performance Assay
Description
- Magnetic format allows for easier wash steps
- Undergoes similar validation testing as Quantikine™ ELISA Kits
- Precise and Specific
- Requires a small sample volume (<50 μL)
- Run your assay in just 3-5 hours
- Mass-calibrated standards for consistent results with every new lot of material
- Enough reagents for one 96-well plate
Specifications
Specifications
| Product Type | Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform |
| Quantity | 1 Kit |
| Species | Human |
| Binding Target | IL-5 |
| Bead Type | Magnetic |
| Assay Sensitivity | 0.12pg/mL |
| Assay Range | 0.34 - 1400pg/mL |
| Gene ID (Entrez) | 3567 |
| Gene Symbol | IL5 |
| Sample Type | Serum, Plasma (EDTA, Heparin) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?