missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-5 Uncoated ELISA Kit with Plates

Product Code. 15541027
Click to view available options
Quantity:
10 x 96 Tests
2 x 96 Tests
20 x 96 Tests
Unit Size:
Pack of 10
Pack of 2
Pack of 20
This item is not returnable. View return policy

Product Code. 15541027

Brand: Invitrogen™ 88705622

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The Human Interleukin-5 (IL-5) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of Human IL-5. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

IL-5 is a cytokine that acts as a growth and differentiation factor for both B cells and eosinophils. IL-5 cytokine is a main regulator of eosinopoiesis, eosinophil maturation and activation. The elevated production of this cytokine is reported to be related to asthma or hypereosinophilic syndromes. The receptor of IL-5 cytokine is a heterodimer, whose beta subunit is shared with the receptors for interleukine 3 (IL3) and colony stimulating factor 2 (CSF2/GM-CSF). IL-5, together with those for interleukin 4 (IL4), interleukin 13 (IL13), and CSF2, form a cytokine gene cluster on chromosome 5. IL-5, IL4, and IL13 are found to be regulated coordinately by long-range regulatory elements spread over 120 kilobases on chromosome 5q31.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05113
Assay Range 4-500 pg/mL
Assay Sensitivity 4 pg/mL
Content And Storage Capture Antibody: Pre-titrated, purified antibody
Detection Antibody: Pre-titrated, biotin-conjugated antibody
Standard: Recombinant protein for generating standard curve and calibrating samples
10X Coating Buffer: Buffer for plating the Capture Antibody
5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme
Enzyme: Pre-titrated Avidin-HRP
Substrate: 1X TMB Solution
Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards
96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22, -76, -86)
Conjugate HRP
Immunoassay Kit Format Uncoated ELISA Kit with Plates
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias B-cell differentiation factor I; B-cell growth factor II; BCGF-II; colony-stimulating factor, eosinophil; cytotoxic T-lymphocyte inducer; EDF; Eosinophil differentiation factor; IL5; IL-5; ILN; Interleukin; Interleukin 5; Interleukin 5 (colony-stimulating factor eosinophil); Interleukin 5 (colony-stimulating factor, eosinophil); Interleukin5; interleukin-5; T-cell replacing factor; TRFB-cell differentiation factor I; B-cell growth factor II; BCGF-II; colony-stimulating factor, eosinophil; cytotoxic T-lymphocyte inducer; EDF; Eosinophil differentiation factor; IL5; IL-5; ILN; Interleukin; Interleukin 5; Interleukin 5 (colony-stimulating factor eosinophil); Interleukin 5 (colony-stimulating factor, eosinophil); Interleukin5; interleukin-5; T-cell replacing factor; TRF;
Gene ID (Entrez) 3567
Gene Symbol Il5
Kit Contents Capture Antibody: Pre-titrated, purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86), Purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86)
Label or Dye Biotin
Quantity 2 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 100 μL, Serum 100 μL, Supernatant 100 μL
Storage Requirements 2-8°C
Synonym Interleukin-5
Target IL-5
Test Time 1 hr 45 min
Total Assay Time 24 hr 30 min
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.