Learn More
Novus Biologicals™ Human IL-4R alpha (Research Grade Dupilumab Biosimilar) Antibody, Novus Biologicals™
GREENER_CHOICE
Human Monoclonal Antibody
Brand:  Novus Biologicals™ MAB11481-100
 View more versions of this product
Product Code. 18714364
  • £491.00 / 100µg
Description
IL-4R alpha Monoclonal antibody specifically detects IL-4R alpha in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-4R alpha
Monoclonal
Unconjugated
P24394
Human
Protein A or G purified from ascites
RUO
3566
Human
Purified
ELISA
Hu209
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
CD124, CD124 antigen, IL-4 R alpha, IL-4 receptor subunit alpha, IL4R, IL4R alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-4 R, Gly24-His232, Accession # P24394
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Use a manual defrost freezer and avoid repeated freeze/thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution.
IgG1