missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-4 Receptor alpha (soluble) Recombinant Protein, PeproTech®

Product Code. 17890583
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 17890583

Brand: Gibco™ 20004RC250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

PeproTech's CHO cell-derived Recombinant Human sIL-4 Receptor alpha is a 23.9 kDa glycoprotein corresponding to 209 amino acid residues of the extracellular domain of IL-4Ralpha. As a result of glycosylation, Recombinant Human sIL-4 Receptor alpha migrates with an apparent molecular mass of approximately 50-65 kDa by SDS-PAGE gel, under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL4 Receptor alpha (IL4RA) is the alpha chain of the interleukin 4 receptor which binds to both interleukin 4 and interleukin 13 to regulate IgE production, chemokine and mucus production at sites of allergic inflammmation. The IL4 response is involved in promoting Th2 differentiation. The secreted extracellular domain of IL4R alpha, called sIL4R alpha, can inhibit IL4-mediated cell proliferation and IL5 upregulation by T-cells. It has no signaling abilities. Allelic variations in the IL4RA gene have been associated with atopy, a condition that can manifest itself as allergic rhinitis, sinusitus, asthma, or eczema.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P24394
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 3566
Molecular Weight (g/mol) 23.9 kDa
Name Human IL-4 Receptor alpha (soluble)
Quantity 250 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 582J2.1; CD124; il-4 receptor; IL-4 receptor alpha chain; IL-4 receptor subunit alpha; IL-4-binding protein; IL4-BP; Il4r; IL4R nirs variant 1; IL-4 R subunit alpha; IL4RA; IL-4 RA; IL-4 R-alpha; interleukin 4 receptor; interleukin 4 receptor, alpha; interleukin-4 receptor alpha chain; interleukin-4 receptor subunit alpha; Soluble IL-4 receptor subunit alpha; Soluble interleukin-4 receptor subunit alpha
Common Name IL4R
Gene Symbol IL4R
Biological Activity The ED50 was determined by its ability to inhibit the IL-4 dependent proliferation of human TF-1 cells is ≤ 5.0 ng/ml (in the presence of 0.5 ng/ml of IL-4), corresponding to a specific activity of ≥ 2 x 10^5 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence GNMKVLQEPT CVSDYMSIST CEWKMNGPTN CSTELRLLYQ LVFLLSEAHT CIPENNGGAG CVCHLLMDDV VSADNYTLDL WAGQQLLWKG SFKPSEHVKP RAPGNLTVHT NVSDTLLLTW SNPYPPDNYL YNHLTYAVNI WSENDPADFR IYNVTYLEPS LRIAASTLKS GISYRARVRA WAQCYNTTWS EWSPSTKWHN SYREPFEQH
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.