missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB304-100
This item is not returnable.
View return policy
Description
IL-4 Monoclonal specifically detects IL-4 in Human samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.
Specifications
| IL-4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3565 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Neutralization, Immunocytochemistry | |
| 3007 | |
| Western Blot 1 ug/mL, Neutralization 0.03-0.1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P05112.1 | |
| IL4 | |
| E. coli-derived recombinant human IL-4 His25-Ser153 Accession # P05112.1 | |
| 100 μg | |
| Primary | |
| Detects human IL-4 in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse IL-4. | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction