missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-3R alpha/CD123 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 7 publications
£180.00 - £1860.00
Specifications
| Antigen | IL-3R alpha/CD123 |
|---|---|
| Clone | 32703 |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, Neutralization 0.6-1.2 ug/mL, Immunocytochemistry 8-25 ug/mL, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18772417
![]() |
R&D Systems
MAB301-SP |
25 μg |
£180.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18151561
![]() |
R&D Systems
MAB301-100 |
100 μg |
£477.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18131251
![]() |
R&D Systems
MAB301-500 |
500 μg |
£1860.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-3R alpha/CD123 Monoclonal specifically detects IL-3R alpha/CD123 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF-ready.Specifications
| IL-3R alpha/CD123 | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, Neutralization 0.6-1.2 ug/mL, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| P26951 | |
| 3563 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-3 R alpha/CD123 Lys20-Arg305, predicted Accession # P26951 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-3 R alpha/CD123 in direct ELISAs and Western blots. |
| 32703 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 R alpha, IL-3 receptor subunit alpha, IL3R, IL3R alpha, IL-3R subunit alpha, IL3RA, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174 | |
| IL3RA | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title