missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-3R alpha/CD123 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF841
Additional Details : Weight : 0.00970kg
Description
IL-3R alpha/CD123 Polyclonal specifically detects IL-3R alpha/CD123 in Human samples. It is validated for Western Blot.Specifications
IL-3R alpha/CD123 | |
Polyclonal | |
Unconjugated | |
CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 R alpha, IL-3 receptor subunit alpha, IL3R, IL3R alpha, IL-3R subunit alpha, IL3RA, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174 | |
Goat | |
Affinity Purified | |
RUO | |
3563 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL | |
IL3RA | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-3 R alpha/CD123 | |
100 μg | |
Primary | |
Detects human IL-3 R alpha/CD123 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 2% cross-reactivity with recombinant human (rh) IL-2 sR beta , rhIL-2 sR gamma , and rhIL-6 sR is observed. | |
Human | |
IgG |
For Research Use Only