missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-38/IL-1F10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB24271-SP
This item is not returnable.
View return policy
Description
IL-38/IL-1F10 Monoclonal specifically detects IL-38/IL-1F10 in Human samples. It is validated for ELISA.
Specifications
| IL-38/IL-1F10 | |
| Monoclonal | |
| Unconjugated | |
| Q8WWZ1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 84639 | |
| Human | |
| Purified |
| ELISA | |
| 892257 | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| FIL1- theta, FKSG75, IL1F10, IL-1HY2, IL38, interleukin 1 family, member 10 (theta), UNQ6119/PRO20041 | |
| E. coli-derived human IL-38/IL-1F10, Ser3-Trp152, Accession # Q8WWZ1 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction