missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Human IL-37 (IL-1F7) ELISA Ready-SET-Go!™ Kit
Click to view available options
Quantity:
2 x 96 Tests
Unit Size:
Pack of 2
Description
Includes
- Capture Antibody Concentrate (250 x)
- Detection Antibody Concentrate (250 x)
- human IL-37 (IL-1F7) standard, (lyophilized), 4000pg/mL upon reconstitution
- Phosphate Buffered Saline Concentrate (PBS) 10x
- Assay Buffer A Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
- Sample Diluent Z
- Tetramethylbenzidine (TMB) Substrate Solution
- 2 96-well plates
Human IL-37 (IL-1F7) Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
Specifications
Specifications
| Assay Sensitivity | 31.3pg/mL |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Quantity | 2 x 96 Tests |
| Synonym | Interleukin-37, IL-1H4 |
| Target | IL-37 (IL-1F7) |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction