missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-37/IL-1F7 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF1975
This item is not returnable.
View return policy
Description
IL-37/IL-1F7 Polyclonal specifically detects IL-37/IL-1F7 in Human samples. It is validated for Western Blot.
Specifications
| IL-37/IL-1F7 | |
| Polyclonal | |
| Biotin | |
| NP_775297 | |
| IL37 | |
| E. coli-derived recombinant human IL-37/IL-1F7 Lys27-Asp192 Accession # NP_775297 | |
| 50 μg | |
| Primary | |
| Detects human IL-37/IL-1F7 in Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human (rh) FIL1 delta is observed and less than 2% cross-reactivity with rhFIL1 epsilon , rhFIL1 eta , rhIL-1 alpha , rhIL-1 beta , and rhIL-1 H1 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| FIL1, FIL1 zeta, FIL1(ZETA), FIL1ZFIL1 zeta, IL-1F7, IL-1F7IL-1 zeta, IL1H4IL1F7 (canonical product IL-1F7b), IL-1H4IL-1F7b (IL-1H4, IL-1H, IL-1RP1), IL-1RP1IL-1X protein, IL1RP1interleukin 1 family member 7, IL-1X, IL37, interleukin 1 family, member 7 (zeta), interleukin 1, zeta, interleukin-1 family member 7, Interleukin-1 homolog 4, interleukin-1 superfamily z, Interleukin-1 zeta, Interleukin-1-related protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 27178 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction