missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-36Ra/IL-1F5 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
£167.00 - £501.00
Specifications
| Antigen | IL-36Ra/IL-1F5 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction 100 ug/mL |
| Applications | Western Blot, Blockade of Receptor-ligand Interaction |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18748617
![]() |
R&D Systems
AF1275-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18062982
|
R&D Systems
AF1275 |
100 μg |
£501.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-36Ra/IL-1F5 Polyclonal specifically detects IL-36Ra/IL-1F5 in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.Specifications
| IL-36Ra/IL-1F5 | |
| Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction 100 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| FIL1, FIL1 delta, FIL1-delta, IL-1RP3, IL-1L1, IL-1-delta), FIL1DIL-1 delta, IL1F5, IL1F5 (Canonical product IL-1F5a), IL-1HY1, IL-1L1, IL-1RP3, IL36RA, IL-36RA, IL36RN, interleukin 1 family, member 5 (delta), interleukin 1, delta, Interleukin 36 Receptor Antagonist, interleukin-1 family member 5, Interleukin-1 HY1, Interleukin-1 receptor antagonist homolog 1, Interleukin-1-like protein 1, Interleukin-36 Receptor Antagonist | |
| IL36RN | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| Unconjugated | |
| RUO | |
| Q9UBH0 | |
| 26525 | |
| E. coli-derived recombinant human IL-36Ra/IL-1F5 Val2-Asp155 Accession # Q9UBH0 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human IL-36Ra/IL-1F5 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) IL-1ra is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title