missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-36Ra/IL-1F5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1275
This item is not returnable.
View return policy
Description
IL-36Ra/IL-1F5 Monoclonal specifically detects IL-36Ra/IL-1F5 in Human samples. It is validated for Western Blot.
Specifications
| IL-36Ra/IL-1F5 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9UBH0 | |
| IL36RN | |
| E. coli-derived recombinant human IL-36Ra/IL-1F5 Met1-Asp155 Accession # Q9UBH0 | |
| 500 μg | |
| Primary | |
| Detects human IL-36Ra/IL-1F5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) IL-1 alpha , rhIL-1 beta , rhIL-1ra, rhIL-18, rhIL-1F6, or rhIL-1F8. | |
| Human | |
| Purified |
| Western Blot | |
| 190524 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FIL1, FIL1 delta, FIL1-delta, IL-1RP3, IL-1L1, IL-1-delta), FIL1DIL-1 delta, IL1F5, IL1F5 (Canonical product IL-1F5a), IL-1HY1, IL-1L1, IL-1RP3, IL36RA, IL-36RA, IL36RN, interleukin 1 family, member 5 (delta), interleukin 1, delta, Interleukin 36 Receptor Antagonist, interleukin-1 family member 5, Interleukin-1 HY1, Interleukin-1 receptor antagonist homolog 1, Interleukin-1-like protein 1, Interleukin-36 Receptor Antagonist | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 26525 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction