missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-36 gamma/IL-1F9 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB2320-500
This item is not returnable.
View return policy
Description
IL-36 gamma/IL-1F9 Monoclonal specifically detects IL-36 gamma/IL-1F9 in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
Specifications
| IL-36 gamma/IL-1F9 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL-1 epsilon, IL-1 H1, IL-1 Related Protein 2, IL-1(EPSILON), IL1E, IL-1-epsilon, IL1F9, IL-1F9, IL1H1, IL-1H1, IL-1-Related Protein 2, IL1RP2, IL-1RP2, IL36 gamma, IL36G, interleukin 1 family, member 9, interleukin 1-related protein 2, Interleukin 36, Gamma, Interleukin-1 epsilon, interleukin-1 family member 9, Interleukin-1 homolog 1, Interleukin-36 Gamma | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 56300 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| 278706 | |
| Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction 0.15-0.6 ug/mL | |
| Q9NZH8 | |
| IL36G | |
| E. coli-derived recombinant human IL-36 gamma/IL-1F9 Met1-Asp169 Accession # Q9NZH8 | |
| 500 μg | |
| Primary | |
| Detects human IL-36 gamma/IL-1F9 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) IL-1 alpha , rhIL-1 beta , rhIL-36Ra/IL-1F5, rhIL-36 alpha/IL-1F6, rhIL-1F7, rhIL-36 beta/IL-1F8, rhIL-1ra/IL-1F3, or rhIL-18/IL-1F4 is observed. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction