missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-36 beta/IL-1F8 DuoSet ELISA
Click to view available options
Format:
96-well Strip Plate
Unit Size:
Pack of 5
Description
Includes
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
The Human IL-36 beta/IL-1F8 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-36 beta/IL-1F8.
Specifications
Specifications
| Product Type | ELISA Kit |
| Assay Sensitivity | 50 ng/mL |
| Cross Reactivity | Not observed |
| Format | 96-well Strip Plate |
| For Use With (Application) | ELISA |
| Sample Type | Cell Culture Supernatant, Serum, Plasma |
| Kit Contents | Capture antibody, Detection antibody, Recombinant standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction