missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-36 alpha (IL-1F6) Recombinant Protein, PeproTech®

Product Code. p-7246164
Click to view available options
Quantity:
10 μg
100 μg
2 μg
2 x 500 μg
250 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
Pack of 2
This item is not returnable. View return policy

Product Code. 17861723

Brand: Gibco™ 20036A100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

200-36A-1MG will be provided as 2 x 500 μg (200-36A-500UG). Recombinant Human IL-36alpha is a 17.2 kDa protein containing 154 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The IL-1 family is comprised of 11 structurally related ligands, including recently re-named IL-36alpha (IL-1F6), beta (IL-1F8), and gamma (IL-1F9). IL-36alpha is highly expressed in psoriatic plaques, as are IL-36beta and IL-36gamma, and is the only IL-1 family member found to be expressed by T cells, as well as monocytes and B cells. Primarily found in skin and lymphoid tissues, IL-36alpha is also found to a lesser extent in tissues of the fetal brain, trachea, stomach, and intestine. IL-36alpha signals through a combination of the IL-1Rrp2 (IL-1R6) receptor, which is primarily expressed on certain dendritic cells, and the widely expressed IL-RAcP (IL-1 receptor accessory protein). IL-36alpha also functions as an agonist of NF-kappaB and MAPK pathways, and induces the production of proinflammatory cytokines and chemokines, including IL-6, IL-8, BD-2, and BD-3.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UHA7
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 27179
Molecular Weight (g/mol) 17.2 kDa
Name Human IL-36 alpha (IL-1F6)
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias FIL1; FIL1 epsilon; FIL1(EPSILON); Fil1e; Fil1epsilon; IL-1 epsilon; IL1(EPSILON); Il1e; Il1f6; IL-1F6; IL-1F6 (FIL-1-epsilon); Il1f9; Il1h1; IL-1H1; IL1RP2; Il36a; Interleukin; interleukin 1; interleukin 1 family, member 6; interleukin 1 family, member 6 (epsilon); interleukin 1 family, member 9; interleukin 1, epsilon; interleukin 36, alpha; interleukin-1 epsilon; Interleukin-1 family member 6; interleukin-1 homolog 1; Interleukin-36; Interleukin-36 alpha
Common Name IL1F6
Gene Symbol IL36A
Biological Activity Measured by its ability to induce IL-8 secretion by A431 human epidermoid carcinoma cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MKIDTPQQGS IQDINHRVWV LQDQTLIAVP RKDRMSPVTI ALISCRHVET LEKDRGNPIY LGLNGLNLCL MCAKVGDQPT LQLKEKDIMD LYNQPEPVKS FLFYHSQSGR NSTFESVAFP GWFIAVSSEG GCPLILTQEL GKANTTDFGL TMLF
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.