missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IL-34 ELISA Kit
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
Pack of 10
Description
IL-34 is secreted as a dimeric protein and currently belongs to no known cytokine family. It signals via the M-CSF Receptor, also known as CSF-1R and CD115, which is shared with M-CSF. IL-34 has no amino acid sequence homology to M-CSF, but they exhibit similarities in secondary structure and share overlapping functions. IL-34 has been shown to promote myeloid survival, proliferation, and differentiation, and to play a role in osteoclast formation. IL-34 expression has been detected in various tissues and is most abundant in the spleen.
Specifications
Specifications
| Accession Number | Q6ZMJ4 |
| Assay Range | 7 to 250 pg/mL |
| Assay Sensitivity | 7 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 146433 |
| Gene Symbol | Il34 |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction