missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-34 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB52651
This item is not returnable.
View return policy
Description
IL-34 Monoclonal specifically detects IL-34 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| IL-34 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C16orf77, IL34, IL-34, interleukin 34 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 146433 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 578410 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q6ZMJ4 | |
| IL34 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-34 Asn21-Pro242 Accession # Q6ZMJ4 | |
| 500 μg | |
| Primary | |
| Detects human IL-34 in sandwich immunoassays. In this format, no cross-reactivity with recombinant mouse IL-34 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction