missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-32 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 11 publications
Brand: R&D Systems AF3040-SP
This item is not returnable.
View return policy
Description
IL-32 Polyclonal specifically detects IL-32 in Human samples. It is validated for Western Blot.
Specifications
| IL-32 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL32, IL-32, IL-32alpha, IL-32beta, interleukin 32, interleukin-32, interleukin-32 theta, natural killer cell transcript 4, Natural killer cells protein 4, NK4, NK4IL-32delta, TAIF, TAIFa, TAIFb, TAIFc, TAIFd, TAIFIL-32gamma, Tumor necrosis factor alpha-inducing factor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9235 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| NP_001012651 | |
| IL32 | |
| E. coli-derived recombinant human IL-32 alpha Cys2-Lys131 Accession # NP_001012651 | |
| 25 μg | |
| Primary | |
| Detects human IL-32 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu