missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-32 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 11 publications
Brand: R&D Systems AF3040
This item is not returnable.
View return policy
Description
IL-32 Polyclonal specifically detects IL-32 in Human samples. It is validated for Western Blot.
Specifications
| IL-32 | |
| Polyclonal | |
| Unconjugated | |
| NP_001012651 | |
| IL32 | |
| E. coli-derived recombinant human IL-32 alpha Cys2-Lys131 Accession # NP_001012651 | |
| 100 μg | |
| Primary | |
| Detects human IL-32 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| IL32, IL-32, IL-32alpha, IL-32beta, interleukin 32, interleukin-32, interleukin-32 theta, natural killer cell transcript 4, Natural killer cells protein 4, NK4, NK4IL-32delta, TAIF, TAIFa, TAIFb, TAIFc, TAIFd, TAIFIL-32gamma, Tumor necrosis factor alpha-inducing factor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9235 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction