missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-32 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB4690-100
This item is not returnable.
View return policy
Description
IL-32 Monoclonal specifically detects IL-32 in Human samples. It is validated for Western Blot.
Specifications
| IL-32 gamma | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| IL32, IL-32, IL-32alpha, IL-32beta, interleukin 32, interleukin-32, interleukin-32 theta, natural killer cell transcript 4, Natural killer cells protein 4, NK4, NK4IL-32delta, TAIF, TAIFa, TAIFb, TAIFc, TAIFd, TAIFIL-32gamma, Tumor necrosis factor alpha-inducing factor | |
| E. coli-derived recombinant human IL-32 gamma protein, Met1-Lys188, Accession # P24001 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
| Western Blot | |
| 2526B | |
| Western Blot 1 ug/mL | |
| P24001 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 9235 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction