missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-32 alpha Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB30401
This item is not returnable.
View return policy
Description
IL-32 alpha Monoclonal specifically detects IL-32 alpha in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| IL-32 alpha | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| NP_001012651 | |
| IL32 | |
| E. coli-derived recombinant human IL-32 alpha Phe3-Lys131 Accession # NP_001012651 | |
| 100 μg | |
| Primary | |
| Detects the alpha isoform of IL-32 in direct ELISAs. In direct ELISAs, less than 15% cross-reactivity with recombinant human (rh) IL-32 beta and rhIL-32 gamma is observed. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 373802 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL32, IL32 alpha, IL-32alpha, IL-32beta, IL-32delta, IL-32gamma, interleukin 32, NK4, TAIF, TAIFa, TAIFb, TAIFc, TAIFd | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9235 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction