missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-32 alpha Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB30401-SP
This item is not returnable.
View return policy
Description
IL-32 alpha Monoclonal specifically detects IL-32 alpha in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| IL-32 alpha | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| NP_001012651 | |
| IL32 | |
| E. coli-derived recombinant human IL-32 alpha Phe3-Lys131 Accession # NP_001012651 | |
| 25 μg | |
| Primary | |
| Detects the alpha isoform of IL-32 in direct ELISAs. In direct ELISAs, less than 15% cross-reactivity with recombinant human (rh) IL-32 beta and rhIL-32 gamma is observed. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 373802 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL32, IL32 alpha, IL-32alpha, IL-32beta, IL-32delta, IL-32gamma, interleukin 32, NK4, TAIF, TAIFa, TAIFb, TAIFc, TAIFd | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9235 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction