missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-31RA Antibody, R&D Systems™
Rat Monoclonal Antibody
£168.00 - £345.00
Specifications
| Antigen | IL-31RA |
|---|---|
| Clone | 313308 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18110041
|
R&D Systems
MAB2769-100 |
100 ug |
£345.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18193471
|
R&D Systems
MAB2769-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-31RA Monoclonal specifically detects IL-31RA in Human samples. It is validated for Flow Cytometry.Specifications
| IL-31RA | |
| Flow Cytometry | |
| Unconjugated | |
| Rat | |
| NP_620586 | |
| 133396 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human IL-31 RA in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse IL-31 RA is observed. |
| 313308 | |
| Monoclonal | |
| Supernatant | |
| Human | |
| class I cytokine receptor, CRL3, CRL3Glmr, Cytokine receptor-like 3, GLMR, GLM-R, GLM-RMGC125346, GP130 like receptor, Gp130-like monocyte receptor, Gp130-like receptor, hGLM-R, IL-31 RA, IL-31 receptor subunit alpha, IL-31R subunit alpha, IL31RA, IL-31RAGPLCRL, IL-31R-alpha, interleukin 31 receptor A, interleukin-31 receptor subunit alpha, PRO21384, soluble type I cytokine receptor CRL3, ZcytoR17 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title