missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-31RA Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB2769S-100UG
This item is not returnable.
View return policy
Description
IL-31RA Monoclonal specifically detects IL-31RA in Human samples. It is validated for Flow Cytometry.
Specifications
| IL-31RA | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| class I cytokine receptor, CRL3, CRL3Glmr, Cytokine receptor-like 3, GLMR, GLM-R, GLM-RMGC125346, GP130 like receptor, Gp130-like monocyte receptor, Gp130-like receptor, hGLM-R, IL-31 RA, IL-31 receptor subunit alpha, IL-31R subunit alpha, IL31RA, IL-31RAGPLCRL, IL-31R-alpha, interleukin 31 receptor A, interleukin-31 receptor subunit alpha, PRO21384, soluble type I cytokine receptor CRL3, ZcytoR17 | |
| NS0-derived rhIL-31 RA extracellular domain aa 20-516 Accession # NP_620586 | |
| 100 μg | |
| Primary | |
| Detects human IL-31 RA in direct ELISAs and western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse IL-31 RA is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry | |
| 313308 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_620586 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 133396 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction