missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-27 R alpha/WSX-1/TCCR APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication
£274.00 - £531.00
Specifications
| Antigen | IL-27R alpha/WSX-1/TCCR |
|---|---|
| Clone | 191106 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18735347
![]() |
R&D Systems
FAB14791A-025 |
25 Tests |
£274.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18756327
![]() |
R&D Systems
FAB14791A-100 |
100 Tests |
£531.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-27R alpha/WSX-1/TCCR Monoclonal specifically detects IL-27R alpha/WSX-1/TCCR in Human samples. It is validated for Flow Cytometry.Specifications
| IL-27R alpha/WSX-1/TCCR | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| class I cytokine receptor, CRL1IL-27R, Cytokine receptor-like 1, IL-27 R alpha, IL27R, IL27R alpha, IL27RA, IL-27RA, IL-27R-alpha, interleukin 27 receptor, alpha, interleukin-27 receptor subunit alpha, TCCR, TCCRIL-27 receptor subunit alpha, T-cell cytokine receptor type 1, Type I T-cell cytokine receptor, WSX-1, WSX1IL-27R subunit alpha, zcytor1 | |
| IL27RA | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human IL-27 R alpha/WSX-1/TCCR in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human gp130 or recombinant mouse IL-27 Ra is observed. |
| 191106 | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| Human | |
| Q6UWB1 | |
| 9466 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-27 R alpha/WSX-1/TCCR Gly34-Lys516 Accession # Q6UWB1 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title