missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-27/IL-35 EBI3 Subunit Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC6456S-100UG
This item is not returnable.
View return policy
Description
EBI3 Monoclonal specifically detects EBI3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-27/IL-35 EBI3 Subunit | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q14213 | |
| EBI3 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-27/IL-35 EBI3 Subunit Arg21-Lys229 (predicted) Accession # Q14213 | |
| RUO | |
| 10148 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 607201 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| cytokine receptor, EBI-3, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 beta, IL-27 subunit beta, IL27B, IL-27B, IL35 EBI3, IL-35 EBI3, interleukin-27 subunit beta | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human IL-27/IL-35 EBI3 Subunit in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) IL-27 EBI3 or rmIL-27 (EBI3 + p28 heterodimer) is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction