missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-26/AK155 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
£171.00 - £1790.00
Specifications
| Antigen | IL-26/AK155 |
|---|---|
| Clone | 197505 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18735499
![]() |
R&D Systems
MAB1375-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18773407
![]() |
R&D Systems
MAB1375-100 |
100 μg |
£462.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18758389
![]() |
R&D Systems
MAB1375-500 |
500 μg |
£1790.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-26/AK155 Monoclonal specifically detects IL-26/AK155 in Human samples. It is validated for Western Blot.Specifications
| IL-26/AK155 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AK155, AK155IL-26interleukin-26, IL26, interleukin 26, Protein AK155 | |
| IL26 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 197505 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q9NPH9 | |
| 55801 | |
| E. coli-derived recombinant human IL-26 Lys22-Gln171 Accession # Q9NPH9 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-26 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human IL-10. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title