missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-24 Recombinant Protein, PeproTech®

Product Code. 17881113
Change view
Click to view available options
Quantity:
100 μg
2 x 500 μg
20 μg
250 μg
5 μg
50 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
50µg
5µg
Pack of 2
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17881113 50 μg 50µg
17871823 100 μg 100µg
17811313 2 x 500 μg 1mg
30088436 2 x 500 μg Pack of 2
17870073 20 μg 20µg
17860343 250 μg 250µg
17830843 500 μg 500µg
17851103 5 μg 5µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 17881113 Supplier Gibco™ Supplier No. 2003550UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

200-35-1MG will be provided as 2 x 500 μg (200-35-500UG). Recombinant Human IL-24 is an 18.9 kDa glycoprotein that contains 161 amino acid residues, including a C-terminal His-tag. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interleukin 24 (IL-24)/melanoma differentiation-associated gene-7 (mda-7) encodes a secreted protein that exhibits significant homology to the interleukin 10 (IL-10) family of cytokines. The human IL-24 is secreted by activated peripheral blood mononuclear cells and binds two heterodimeric receptors, IL-22R1/IL-20R2 and IL-20R1/IL-20R2 resulting in the activation of JAK/STAT signaling pathways. Mda-7/IL-24, selectively induces growth suppression, apoptosis and radiosensitization in malignant gliomas in a p53-independent manner.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13007
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 11009
Molecular Weight (g/mol) 18.9 kDa
Name Human IL-24
Quantity 50 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias C49A; Cytokine-like protein Mob-5; FISP; IL 24; IL10B; Il24; IL-24; IL-4-induced secreted protein; Interleukin; interleukin 24; interleukin-24; Mda7; MDA-7; melanocyte-associated Mda-7; melanoma differentiation-associated gene 7 protein; Mob5; ST16; suppression of tumorigenicity 16 (melanoma differentiation); Suppression of tumorigenicity 16 protein; Th2-specific cytokine FISP
Common Name IL24
Gene Symbol IL24
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence QEFHFGPCQV KGVVPQKLWE AFWAVKDTMQ AQDNITSARL LQQEVLQNVS DAESCYLVHT LLEFYLKTVF KNYHNRTVEV RTLKSFSTLA NNFVLIVSQL QPSQENEMFS IRDSAHRRFL LFRRAFKQLD VEAALTKALG EVDILLTWMQ KFYKLHHHHH H
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.