missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-24 Recombinant Protein, PeproTech®

Product Code. 17860343
Click to view available options
Quantity:
100 μg
2 x 500 μg
20 μg
250 μg
5 μg
50 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
50µg
5µg
Pack of 2
This item is not returnable. View return policy

Product Code. 17860343

Brand: Gibco™ 20035250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

200-35-1MG will be provided as 2 x 500 μg (200-35-500UG). Recombinant Human IL-24 is an 18.9 kDa glycoprotein that contains 161 amino acid residues, including a C-terminal His-tag. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interleukin 24 (IL-24)/melanoma differentiation-associated gene-7 (mda-7) encodes a secreted protein that exhibits significant homology to the interleukin 10 (IL-10) family of cytokines. The human IL-24 is secreted by activated peripheral blood mononuclear cells and binds two heterodimeric receptors, IL-22R1/IL-20R2 and IL-20R1/IL-20R2 resulting in the activation of JAK/STAT signaling pathways. Mda-7/IL-24, selectively induces growth suppression, apoptosis and radiosensitization in malignant gliomas in a p53-independent manner.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13007
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 11009
Molecular Weight (g/mol) 18.9 kDa
Name Human IL-24
Quantity 250 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias C49A; Cytokine-like protein Mob-5; FISP; IL 24; IL10B; Il24; IL-24; IL-4-induced secreted protein; Interleukin; interleukin 24; interleukin-24; Mda7; MDA-7; melanocyte-associated Mda-7; melanoma differentiation-associated gene 7 protein; Mob5; ST16; suppression of tumorigenicity 16 (melanoma differentiation); Suppression of tumorigenicity 16 protein; Th2-specific cytokine FISP
Common Name IL24
Gene Symbol IL24
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence QEFHFGPCQV KGVVPQKLWE AFWAVKDTMQ AQDNITSARL LQQEVLQNVS DAESCYLVHT LLEFYLKTVF KNYHNRTVEV RTLKSFSTLA NNFVLIVSQL QPSQENEMFS IRDSAHRRFL LFRRAFKQLD VEAALTKALG EVDILLTWMQ KFYKLHHHHH H
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.