missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IL-24 ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
Description
Human IL-24 quantitates human IL-24 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human IL-24.
Interleukin 24 (IL-24)/melanoma differentiation-associated gene-7 (mda-7) encodes a secreted protein that exhibits significant homology to the interleukin 10 (IL-10) family of cytokines. The human IL-24 is secreted by activated peripheral blood mononuclear cells and binds two heterodimeric receptors, IL-22R1/IL-20R2 and IL-20R1/IL-20R2 resulting in the activation of JAK/STAT signaling pathways. Mda-7/IL-24, selectively induces growth suppression, apoptosis and radiosensitization in malignant gliomas in a p53-independent manner.
Specifications
Specifications
| Accession Number | C49A; Cytokine-like protein Mob-5; FISP; IL 24; IL10B; Il24; IL-24; IL-4-induced secreted protein; interleukin 24; interleukin-24; MDA7; MDA-7; melanocyte-associated Mda-7; melanoma differentiation-associated gene 7 protein; Mob5; St16; suppression of tumorigenicity 16 (melanoma differentiation); Suppression of tumorigenicity 16 protein; Th2-specific cytokine FISP |
| Assay Range | 0.41 to 100 ng/mL |
| Assay Sensitivity | 0.41 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 11009 |
| Gene Symbol | IL24 |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction