missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-23R PerCP-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB14001C-025
This item is not returnable.
View return policy
Description
IL-23R Monoclonal specifically detects IL-23R in Human samples. It is validated for Flow Cytometry.
Specifications
| IL-23R | |
| Monoclonal | |
| PerCP | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| IBD17, IL-23 R, IL-23 receptor, IL23R, IL-23R, interleukin 23 receptor, interleukin-23 receptor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 149233 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 218213 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Q5VWK5 | |
| IL23R | |
| Mouse myeloma cell line NS0-derived recombinant human IL-23 R Gly24-Ile354 Accession # Q5VWK5 | |
| 25 Tests | |
| Primary | |
| Detects human IL-23 R in direct ELISAs and Western blots. Does not cross-react with recombinant mouse IL-23 R. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction