missing translation for 'onlineSavingsMsg'
Learn More

Human IL-23, Invitrogen™

Product Code. 15238994
Click to view available options
Quantity:
10 μg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15238994

Brand: Invitrogen 14823963

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

IL-23 is a heterodimeric cytokine composed of the p40 subunit of IL-12 disulfide-linked with a protein p19. p19, like p35 of IL-12, is biologically inactive by itself. IL-23 interacts with IL-12Rbeta1 and an additional, novel beta2-like receptor subunit with STAT4 binding domain, termed IL-23R. IL-23 is secreted by activated mouse and human dendritic cells. Biological activities of mouse IL-23 are distinct from those of mouse IL-12. Mouse IL-23 was found not to induce significant amounts of IFN-gamma. Mouse IL-23 does induce strong proliferation of memory T cells (but not naive T cells), whereas IL-12 has no effect on memory cells. Additionally, mouse IL-23 (but not IL-12) can activate mouse memory T cells to produce the proinflammatory cytokine IL-17. Human IL-23 has biological properties which are less distinct from human IL-12; human IL-23 induces proliferation of memory T cells and induces moderate levels of IFN-gamma production by naive and memory T cells, as compared to IL-12. eBioscience′s recombinant IL-23 is produced in baculovirus-infected insect cells as an authentic heterodimer of precursor p19 and p40 subunits using a dual promoter expression system. It is distinct from other available forms of the protein in that it is expressed as a true heterodimer, as opposed to a single-chain, pseudo-heterodimer in which the subunits are joined by an artificial linker. Description: IL-23 is a heterodimeric cytokine composed of the p40 subunit of IL-12 disulfide-linked with a protein p19. p19, like p35 of IL-12, is biologically inactive by itself. IL-23 interacts with IL-12Rbeta1 and an additional, novel beta2-like receptor subunit with STAT4 binding domain, termed IL-23R. IL-23 is secreted by activated mouse and human dendritic cells. Biological activities of mouse IL-23 are distinct from those of mouse IL-12. Mouse IL-23 was found not to induce significant amounts of IFN-gamma. Mouse IL-23 does induce strong proliferation of memory T cells (but not naive T cells), whereas IL-12 has no effect on memory cells. Additionally, mouse IL-23 (but not IL-12) can activate mouse memory T cells to produce the proinflammatory cytokine IL-17. Human IL-23 has biological properties which are less distinct from human IL-12; human IL-23 induces proliferation of memory T cells and induces moderate levels of IFN-gamma production by naive and memory T cells, as compared to IL-12. eBioscience′s recombinant IL-23 is produced in baculovirus-infected insect cells as an authentic heterodimer of precursor p19 and p40 subunits using a dual promoter expression system. It is distinct from other available forms of the protein in that it is expressed as a true heterodimer, as opposed to a single-chain, pseudo-heterodimer in which the subunits are joined by an artificial linker. Applications Reported: Recombinant human IL-23 is biologically active. Applications Tested: The ED50 of this protein, as measured by induction of IL-17A in mouse splenocytes, is less than or equal to 3.5ng/mL. This corresponds to a specific activity of greater than or equal to 2.9 x 10e5 Units/mg. Source: Insect cells infected with baculovirus: human p40, amino acids met 1-Ser 328, accession &Num; NM_002187 was co-expressed with human p19, amino acids met 1-pro 189, accession &Num; NM_016584. Bioactivity: The ED50 of this protein, as measured by induction of IL-17A in mouse splenocytes, is less than or equal to 3.5ng/mL. This corresponds to a specific activity of greater than or equal to 2.9 x 10e5 Units/mg. Endotoxin: Less than 0.01ng/ug cytokine as determined by the LAL assay. Purity: >98% as determined by SDS-PAGE. Molecular Weight: The heterodimer of p40, amino acids ile 23-ser 328, cystine linked to p19, amino acids arg 20-pro 189, has a predicted molecular mass of 53,356. On non-reducing SDS-PAGE the heterodimeric cystine-linked protein migrates as a 55 kDa protein. The DTT reduced protein migrates as 43 kDa and 18 kDa polypeptides. Storage and handling: For best recovery, quick-spin vial prior to opening. Use in a sterile environment. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2μm post-manufacturing filtered.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NPF7
Concentration 0.1mg/mL
For Use With (Application) Control, ELISA, Functional Assay
Formulation 20mM phosphate with 0.2M NaCl, 1% BSA and no preservative; pH 6.0
Gene ID (Entrez) 51561
Molecular Weight (g/mol) The heterodimer of p40, amino acids ile 23-ser 328, cystine linked to p19, amino acids arg 20-pro 189, has a predicted molecular mass of 53,356. On non-reducing SDS-PAGE the heterodimeric cystine-linked protein migrates as a 55kDa protein. The DTT reduced protein migrates as 43kDa and 18kDa polypeptides.
Name Human IL-23
pH Range 6.0
Purification Method SDS-PAGE
Quantity 10 μg
Source Insect cells infected with baculovirus
Storage Requirements -80°C
Regulatory Status RUO
Structural Form Insect cells infected with baculovirus: human p40, amino acids met 1-ser 328, accession # NM_002187 was co-expressed with human p19, amino acids met 1-pro 189, accession # NM_016584
Endotoxin Concentration Less than 0.01ng/μg cytokine as determined by the LAL assay.
Common Name Human IL-23
Gene Symbol IL23A
Biological Activity The ED50 of this protein, as measured by induction of IL-17A in mouse splenocytes, is less than or equal to 3.5ng/mL. This corresponds to a specific activity of greater than or equal to 2.9 x 105 Units/mg.
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species Baculovirus
Recombinant Recombinant
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade Affinity chromatography
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.