Learn More
Gibco™ Human IL-23 Recombinant Protein
Description
Carrier-Free Reconstitution: Lyophilized human IL-23 should be reconstituted in water to a protein concentration of 0.1-1.0 mg/mL. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein, such as 0.1% BSA and store in working aliquots at -80°C to -20°C. Storage: Lyophilized human IL-23 should be stored at 2°C to 8°C, preferably desiccated. Store reconstituted human IL-23 at ≤-20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.
Specifications
Specifications
| Accession Number | Q9NPF7 |
| For Use With (Application) | Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 51561 |
| Molecular Weight (g/mol) | 55 kDa |
| Name | Human IL-23 |
| Purification Method | Purified |
| Quantity | 100 μg |
| Storage Requirements | 4°C |
| Regulatory Status | RUO |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.