missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-23 p19 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB12901-100
This item is not returnable.
View return policy
Description
IL-23A/IL-23 P19 Monoclonal specifically detects IL-23A/IL-23 P19 in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| IL-23A/IL-23 P19 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IL23, IL-23, IL-23 p19, IL-23 p19/IL-12 p40, IL23A, IL-23A, IL-23-A, IL-23p19, IL-23p19/IL-12p40, IL23P19P19, interleukin 23 p19 subunit, interleukin 23, alpha subunit p19, interleukin-23 subunit alpha, Interleukin-23 subunit p19, JKA3 induced upon T-cell activation, MGC79388, SGRF, SGRFIL-23 subunit alpha | |
| E. coli-derived recombinant human IL-23 p19 Arg20-Pro189 Accession # Q9NPF7 | |
| 100 μg | |
| Primary | |
| Detects human IL-23 p19 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Neutralization | |
| 1193B | |
| Western Blot 1 ug/mL, Neutralization 0.15-0.9 ug/mL | |
| Q9NPF7 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 51561 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction