missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-22BP Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF1087-SP
This item is not returnable.
View return policy
Description
IL-22BP/IL22 RA2 Polyclonal specifically detects IL-22BP/IL22 RA2 in Human samples. It is validated for Western Blot.
Specifications
| IL-22BP/IL22 RA2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| class II cytokine receptor, CRF2-10, CRF2-S1IL22BP, CRF2-X, Cytokine receptor class-II member 10, Cytokine receptor family 2 member 10, Cytokine receptor family type 2, soluble 1, IL-22 RA2, IL-22 receptor subunit alpha-2, IL22BP, IL-22BPCRF2X, IL22RA2, IL-22RA2, IL-22R-alpha-2, interleukin 22 receptor, alpha 2, interleukin 22-binding protein, interleukin-22 receptor subunit alpha-2, Interleukin-22-binding protein, MGC150509, MGC150510, zcytoR16 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 116379 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| NP_851826 | |
| IL22RA2 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-22BP Thr22-Pro231 Accession # NP_851826 | |
| 25 μg | |
| Primary | |
| Detects human IL-22BP in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human IL-22 R is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction