missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-21 Human Uncoated ELISA Kit with Plates
Specifically engineered for accurate and precise measurement of IL-21 protein levels from samples including serum, plasma, and cell culture supernatants. This second generation kit has increased sensitivity with a range from 8-1000pg/mL.
£174.00 - £546.00
Specifications
Target Species | Human |
---|---|
For Use With (Application) | ELISA |
Synonym | Interleukin-21 |
Alert:
Contains Sodium azide
Product Code | Brand | Quantity | Gene Symbol | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Gene Symbol | Price | Quantity & Availability | ||||
15501217
|
Invitrogen™
88-8218-76 |
20 x 96 Tests | IL-21 |
£546.00
Pack of 20 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
15511217
|
Invitrogen™
88-8218-77 |
20 x 96 Tests | IL-21 |
£524.00
Pack of 20 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
15591207
|
Invitrogen™
88-8218-22 |
2 x 96 Tests | BSF-1,IL-4 |
£174.00
Pack of 2 |
Estimated Shipment: 20-05-2024
Log in to see stock. |
||||
15521217
|
Invitrogen™
88-8218-86 |
10 x 96 Tests | BSF-1,IL-4 |
£392.00
Pack of 10 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
13582750
|
Invitrogen™
88-8218-88 |
10 x 96 Tests | IL-6 |
£367.00
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
Description
IL-21 Human Uncoated ELISA Kit with Plates contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)Specifications
Human | |
ELISA | |
Interleukin-21 |
For Research Use Only