missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-21 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£171.00 - £428.00
Specifications
| Antigen | IL-21 |
|---|---|
| Clone | 148002 |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18139710
![]() |
R&D Systems
MAB1500-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18791577
![]() |
R&D Systems
MAB1500-100 |
100 μg |
£428.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-21 Monoclonal specifically detects IL-21 in Human samples. It is validated for ELISA.Specifications
| IL-21 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9HBE4 | |
| 59067 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 148002 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CVID11, IL21, IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform, Za11 | |
| E. coli-derived recombinant human IL-21, Gln25-Ser155, Accession # Q9HBE4 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title