missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-20 Quantikine ELISA Kit

Product Code. 18792454
Click to view available options
Sufficient For:
1 x 96-well Plate
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18792454

Brand: R&D Systems™ DL200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human IL-20 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-20 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-20 in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 62.50 - 4000pg/mL
Assay Sensitivity 16.6pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity Natural and Recombinant Human IL-20
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 50604
Sample Volume Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym Cytokine Zcyto10, IL10D, IL20, IL-20 ZCYTO10 four alpha helix cytokine, interleukin 20, interleukin-20, MGC96907
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.