missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-2 Recombinant Protein

Product Code. 15511937
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 15511937

Brand: Invitrogen™ BMS334

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Description: Recombinant human IL-2 produced in E.Coli is a single, non-glycosylated mutein (variant form) of human IL-2 polypeptide chain containing 133 amino acids and having a molecular mass of 15385.79 Dalton. eBioscience's rhIL-2 is Des N-Terminal Met and has an Ala substitute for Cysteine at position 126. BMS334 is fully biologically active when compared to standard. The ED50 as determined by the dose-dependent stimulation of murine CTLL-2 cells is <0.0645 ng/mL, corresponding to a specific activity of 1.55x107 IU/mg. Purity: >99%. Molecular Weight: 15.4 kDa.

Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P60568
For Use With (Application) Neutralization
Formulation Protein with no preservative
Gene ID (Entrez) 3558
Molecular Weight (g/mol) 15.4 kDa
Name Human IL-2
Purification Method Purified
Quantity 50 μg
Source E. coli
Storage Requirements 4°C
Regulatory Status RUO
Gene Alias aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF
Common Name IL-2
Gene Symbol IL2
Biological Activity ED50 < 0.0645 ng/mL; determined by dose-dependent stimulation of murine CTLL-2 cells. This corresponds to a specific activity of 1.55 x 10e7 IU/mg.
Product Type Protein
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Sequence Human IL-2 recombinant protein contains 133 amino acids. It contains and N-terminal Met and a A126C substitution.
Content And Storage 4°C
Activity ED50 < 0.0645 ng/mL; determined by dose-dependent stimulation of murine CTLL-2 cells. This corresponds to a specific activity of 1.55x10e7 IU/mg.
Expression System E. coli
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade >99%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.