missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human IL-2, Recombinant Protein, Carrier-Free

Product Code. 15558736
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
This item is not returnable. View return policy

Product Code. 15558736

Brand: Invitrogen™ 34802985

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Human Recombinant Protein

Human IL-2 (also known as TCGF) is a ∽15 kDa factor produced mainly by activated CD4+ T cells. IL-2 induces cell cycle progression of resting cells in an antigen non-specific manner and allows clonal expansion of activated T cells. IL-2 also acts on activated B cells, monocytes, NK, LAK cells, and on oligodendroglial cells in vitro. In addition, IL-2 plays a role in hematopoiesis, tumor surveillance and anti-inflammatory reactions and hence is a central regulator of the immune response. Non-glycosylated IL-2 is biologically active.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5mg/mL
For Use With (Application) Cytokine Bioassay, ELISA
Formulation Sterile liquid: 40mM acetate, 0.2M NaCl, pH 5. 0.22μm filtered.
Molecular Weight (g/mol) 15kDa
Name IL-2
Purification Method SDS-PAGE
Quantity 500 μg
Source E. coli
Storage Requirements Store at less than or equal to -70°C.
Regulatory Status RUO
Structural Form E. coli
Endotoxin Concentration Less than 0.01ng/μg cytokine as determined by the LAL assay.
Gene Symbol Interleukin-2
Biological Activity The ED50 of this protein, as measured by CTLL-2 proliferation assay, is less than or equal to 175pg/mL. This corresponds to a specific activity of greater than or equal to 5.7 x 106 Units/mg.
Product Type Recombinant Protein, Carrier-Free
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Purity or Quality Grade >98%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.