missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-2 R beta Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF-224-SP
This item is not returnable.
View return policy
Description
IL-2 R beta Polyclonal specifically detects IL-2 R beta in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| IL-2 R beta | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD122, CD122 antigen, high affinity IL-2 receptor beta subunit, High affinity IL-2 receptor subunit beta, IL-15 R beta, IL-2 receptor subunit beta, IL2R beta, IL-2R subunit beta, IL2RB, IL-2RB, interleukin 15 receptor, beta, interleukin 2 receptor, beta, interleukin-2 receptor subunit beta, P70-75, p75 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3560 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 10-30 ug/mL | |
| NP_000869 | |
| IL2RB | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-2 R beta Ala27-Asp239 Accession # NP_000869 | |
| 25 μg | |
| Primary | |
| Detects human IL-2 R beta in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) IL-2 R alpha , recombinant mouse (rm) IL-2 R beta , rhIL-2 R gamma , and rhIL-15 R is observed. Is also able to block the cell surface of human IL-2 R beta mediated bioactivities induced by IL-2. For optimal neutralization of IL-2 biological activity on cells expressing the high affinity IL-2 receptors, the use of anti-IL-2 R alpha in conjunction with anti-IL-2 R beta antibodies is recommended. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction