missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Human IL-2 Platinum ELISA Kit, for in vitro diagnostic use
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
Description
Includes
Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-2
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
Specifications
| Assay Sensitivity | 9.2pg/mL |
| Conjugate | Biotin |
| Sample Type | Amniotic Fluid, Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine, Whole Blood |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Quantity | 96 Tests |
| Sample Volume | 50 μL |
| Synonym | Aldesleukin, T cell growth factor, TCGF Aldesleukin, T cell growth factor, TCGF |
| Target | IL-2 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction