Learn More
Invitrogen™ IL-2 Human Instant ELISA™ Kit
Instant ELISA Kit
Brand: Invitrogen™ BMS221INST
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminum pouch(es) with a Microwell Plate coated with Monoclonal Antibody (murine) to human IL-2, Biotin-Conjugate (anti-IL-2 polyclonal antibody), Sample Diluent and Streptavidin-HRP, lyophilized
Aluminum pouch(es) with a human IL-2 Standard curve (coloured)
Wash Buffer Concentrate 20x (phosphate-buffered saline with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Sample Diluent (Use when an external predilution of the samples is needed)
Stop Solution (1M Phosphoric acid)
Adhesive Plate Covers
Description
The Human Interleukin 2 (Hu IL-2) ELISA quantitates Hu IL-2 in human serum, plasma, cell culture supernatants, amniotic fluid, synovial fluid, urine or other body fluids. The assay will exclusively recognize both natural and recombinant Hu IL-2. Principle of the method The Human IL-2 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.Specifications
P60568 | |
2.3 pg/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
IL-2,TCGF,lymphokine | |
5.8% | |
HRP | |
RUO | |
-20°C | |
Human | |
3 hr. 10 min. |
18.8-1,200 pg/mL | |
Biotin | |
Amniotic Fluid, Plasma, Serum, Supernatant, Synovial Fluid, Urine | |
ELISA | |
3558 | |
7.4% | |
Pre-coated 96 well Microplate, Adhesive Plate Covers, Biotinylated Detection Antibody, Sample Diluent, Sample Diluent, lyophilized, Wash Buffer Concentrate, Wash Buffer Concentrate, Stop Solution, Stop Solution, Lysis Buffer 10x | |
128 Tests | |
Amniotic Fluid, 50 μL; Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL; Synovial Fluid,50 μL; Urine,50 μL | |
Aldesleukin, T cell growth factor, TCGF | |
20 min. |
For Research Use Only