Learn More
R&D Systems™ Human IL-2 ELISpot Development Module, 5 Plate
Product Code. 18412240
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. Quantity unitSize
18412240 1 Kit 1 set
1 options
Product Code. 18412240 Supplier R&D Systems™ Supplier No. SEL202
Sufficient for at least five 96-well plates
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.